Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGDIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001175.6 | NP_001166.3 | 6 | 830 | ||
NM_001321420.1 | NP_001308349.1 | 7 | 988 | |||
NM_001321421.1 | NP_001308350.1 | 6 | 709 | |||
NM_001321422.1 | NP_001308351.1 | 6 | 831 | |||
NM_001321423.1 | NP_001308352.1 | 6 | 697 | |||
NR_135637.1 | 4 | 416 | ||||
GenBank mRNA | AB451315.1 | - | 5 | 584 | ||
AB451445.1 | - | 5 | 584 | |||
AF498927.1 | - | 5 | 584 | |||
AK311807.1 | - | 6 | 660 | |||
BC009200.2 | - | 6 | 634 | 3621138 | ||
BI762648.1 | - | 6 | 828 | |||
BM849963.1 | - | 3 | 278 | |||
BP365648.1 | - | 4 | 416 | |||
BQ213418.1 | - | 1 | 179 | |||
L07916.1 | - | 6 | 658 | |||
L20688.1 | - | 6 | 736 | |||
X69549.1 | - | 6 | 638 |