Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001636.3 | NP_001627.2 | 4 | 1093 | ||
GenBank mRNA | AB209822.1 | - | 4 | 1048 | ||
AK054789.1 | - | 1456 | ||||
AY007135.1 | - | 4 | 1045 | |||
BC007295.1 | - | 4 | 1008 | 3349670 | ||
BC007850.2 | - | 4 | 1047 | 4136545 | ||
BC008737.2 | - | 4 | 1035 | 3342722 | ||
BC008935.2 | - | 4 | 1040 | 2824067 | ||
BC013256.1 | - | 4 | 1047 | |||
BC014775.1 | - | 4 | 1046 | 3458777 | ||
BC031912.1 | - | 4 | 993 | 5141625 | ||
BC035469.1 | - | 4 | 1047 | |||
J03592.1 | - | 3 | 848 |