Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001636.3 | NP_001627.2 | 4 | 1227 | ||
GenBank mRNA | AB209822.1 | - | 4 | 1182 | ||
AK054789.1 | - | 1590 | ||||
AY007135.1 | - | 4 | 1179 | |||
BC007295.1 | - | 4 | 1142 | 3349670 | ||
BC007850.2 | - | 4 | 1181 | 4136545 | ||
BC008737.2 | - | 4 | 1169 | 3342722 | ||
BC013256.1 | - | 4 | 1181 | |||
BC014775.1 | - | 4 | 1180 | 3458777 | ||
BC031912.1 | - | 4 | 1127 | 5141625 | ||
BC035469.1 | - | 4 | 1181 | |||
J03592.1 | - | 3 | 982 |