Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304451.1 | NP_001291380.1 | 3 | 278 | ||
NM_001304452.1 | NP_001291381.1 | 3 | 357 | |||
NM_002631.3 | NP_002622.2 | 3 | 278 | |||
GenBank mRNA | AK290404.1 | - | 3 | 203 | ||
AK298830.1 | - | 3 | 357 | |||
AK300952.1 | - | 3 | 236 | |||
AK303898.1 | - | 3 | 203 | |||
AK304423.1 | - | 3 | 201 | |||
BC000368.2 | - | 3 | 189 | 2819330 | ||
CD700605.1 | - | 3 | 280 | |||
HY049516.1 | - | 3 | 279 | |||
U30255.1 | - | 2 | 158 |