Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002067.4 | NP_002058.2 | 7 | 1538 | ||
GenBank mRNA | AK091639.1 | - | 2 | 2616 | ||
BC089041.1 | - | 7 | 1538 | 6085072 | ||
HY118575.1 | - | 1 | 42 | |||
M69013.1 | - | 7 | 1481 |