Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002073.2 | NP_002064.1 | 2, 3 | 1256 | ||
GenBank mRNA | AF493899.1 | - | 1, 2 | 719 | ||
AK293653.1 | - | 3, 4 | 767 | |||
AK312519.1 | - | 2, 3 | 1197 | |||
BC026342.1 | - | 2, 3 | 1256 | 4814858 | ||
BC037333.1 | - | 2, 3 | 1394 | 5264642 | ||
BC078163.1 | - | 1, 2 | 1094 | 6180193 | ||
BC096828.1 | - | 1, 2 | 722 | 6519151 | ||
CR456495.1 | - | 1, 2 | 806 | |||
J03260.1 | - | 1, 2 | 731 |