Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130841.1 | NP_001124313.1 | 5 | 577 | ||
NM_003729.3 | NP_003720.1 | 4 | 538 | |||
XM_005271298.4 | XP_005271355.1 | 4 | 535 | |||
XM_005271299.4 | XP_005271356.1 | 4 | 524 | |||
XM_017002669.1 | XP_016858158.1 | 4 | 281 | |||
GenBank mRNA | AK292366.1 | - | 4 | 487 | ||
AK299793.1 | - | 4 | 539 | |||
AU310497.1 | - | 4 | 539 | |||
BC012604.1 | - | 5 | 491 | 4281547 | ||
DA361597.1 | - | 4 | 480 | |||
Y11651.1 | - | 4 | 490 |