Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF110532.1 | - | 2 | 231 | ||
AK291427.1 | - | 2 | 439 | |||
AK295233.1 | - | 2 | 492 | |||
AK302906.1 | - | 2 | 449 | |||
AK315940.1 | - | 2 | 420 | |||
AY358711.1 | - | 2 | 257 | |||
BC033091.1 | - | 2 | 482 | 5455903 | ||
DC314392.1 | - | 2 | 509 | |||
DR002070.1 | - | 2 | 431 |