Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204051.1 | NP_001190980.1 | 6 | 912 | ||
NM_001204052.1 | NP_001190981.1 | 6 | 912 | |||
NM_004277.4 | NP_004268.3 | 6 | 912 | |||
XM_005249483.2 | XP_005249540.1 | 6 | 882 | |||
XM_017011512.1 | XP_016867001.1 | 5 | 761 | |||
GenBank mRNA | AF110532.1 | - | 6 | 634 | ||
AK090871.1 | - | 4 | 1241 | |||
AK291427.1 | - | 6 | 842 | |||
AK295233.1 | - | 6 | 895 | |||
AK302906.1 | - | 7 | 952 | |||
AK315940.1 | - | 6 | 823 | |||
AY358711.1 | - | 6 | 660 | |||
BC033091.1 | - | 6 | 885 | 5455903 |