Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242505.2 | NP_001229434.1 | 1, 2 | 327 | ||
NM_001242506.2 | NP_001229435.1 | 1, 2 | 327 | |||
NM_004293.4 | NP_004284.1 | 1, 2 | 327 | |||
XM_005252317.1 | XP_005252374.1 | 1, 2 | 325 | |||
XM_011519213.1 | XP_011517515.1 | 1, 2 | 325 | |||
XM_011519214.1 | XP_011517516.1 | 1, 2 | 325 | |||
XM_011519215.1 | XP_011517517.1 | 1, 2 | 325 | |||
XM_011519216.1 | XP_011517518.1 | 1, 2 | 325 | |||
XM_011519217.1 | XP_011517519.1 | 1, 2 | 325 | |||
XM_011519222.1 | XP_011517524.1 | 1, 2 | 325 | |||
XM_017015333.1 | XP_016870822.1 | 1, 2 | 325 | |||
XM_017015334.1 | XP_016870823.1 | 1, 2 | 325 | |||
XM_017015335.1 | XP_016870824.1 | 1, 2 | 325 | |||
XM_017015336.1 | XP_016870825.1 | 1, 2 | 325 | |||
XM_017015337.1 | XP_016870826.1 | 1, 2 | 325 | |||
GenBank mRNA | AB033084.1 | - | 1, 2 | 194 | ||
AF019638.1 | - | 1, 2 | 203 | |||
AF095286.1 | - | 1, 2 | 211 | |||
AK315988.1 | - | 1, 2 | 105 | |||
BC053584.1 | - | 1, 2 | 301 | 5186337 |