Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGDIA SIRNA Assays ›
Gene Symbol:
ARHGDIA
Gene Name:
Rho GDP dissociation inhibitor alpha
Gene Aliases:
GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1
Chromosome Location:
Chr.17: 81867719 - 81871406 on Build GRCh38
UniGene ID:
Hs.159161
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
119694
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGDIA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001185077.2
NP_001172006.1
4
661
NM_001185078.2
NP_001172007.1
4
431
NM_001301240.1
NP_001288169.1
4
431
NM_001301241.1
NP_001288170.1
4
661
NM_001301242.1
NP_001288171.1
4
431
NM_001301243.1
NP_001288172.1
3
566
NM_004309.5
NP_004300.1
4
431
XM_011523574.1
XP_011521876.1
3
791
GenBank mRNA
AF498926.1
-
3
295
AK293147.1
-
4
375
AK300816.1
-
4
404
AK312347.1
-
4
367
BC005851.2
-
4
387
2961556
BC005875.2
-
4
387
2961556
BC008701.3
-
3
322
2966760
BC009759.2
-
4
352
3935956
BC016031.1
-
3
303
3911525
BC016185.1
-
4
347
4867857
BC024258.1
-
4
347
3027721
BC027730.2
-
4
350
4764249
BC028333.1
-
4
350
BC075827.1
-
4
347
6074977
BC106044.1
-
4
390
6162284
BI755343.1
-
4
352
BI768191.1
-
4
388
BM552638.1
-
4
601
BP325393.1
-
4
362
BQ674863.1
-
4
345
BT006884.1
-
3
295
CF132249.1
-
3
490
CR456777.1
-
3
295
D13989.1
-
3
315
DC406556.1
-
4
431
EU794615.1
-
3
322
KU177916.1
-
3
295
KU177917.1
-
3
295
M97579.1
-
3
295
X69550.1
-
4
348
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
negative regulation of cell adhesion
regulation of Rho protein signal transduction
negative regulation of apoptotic process
positive regulation of GTPase activity
negative regulation of axonogenesis
positive regulation of axonogenesis
regulation of small GTPase mediated signal transduction
semaphorin-plexin signaling pathway
regulation of actin cytoskeleton reorganization
Rho GDP-dissociation inhibitor activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline