Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005469.3 | NP_005460.2 | 4 | 594 | ||
XM_005260239.3 | XP_005260296.1 | 3 | 456 | |||
XM_011528480.2 | XP_011526782.1 | 5 | 618 | |||
GenBank mRNA | AF014404.1 | - | 4 | 530 | ||
AF124264.1 | - | 4 | 500 | |||
AK056668.1 | - | 3 | 420 | |||
AK126206.1 | - | 2 | 673 | |||
AK292494.1 | - | 3 | 422 | |||
AK296973.1 | - | 4 | 582 | |||
AK298783.1 | - | 2 | 194 | |||
BC117155.1 | - | 4 | 522 | 40125706 | ||
BC117157.1 | - | 4 | 522 | 40125708 | ||
BI911038.1 | - | 4 | 580 | |||
CR541771.1 | - | 4 | 500 | |||
CR541778.1 | - | 4 | 500 | |||
X86032.1 | - | 4 | 568 |