Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042537.1 | NP_001036002.1 | 16 | 4042 | ||
NM_001177651.1 | NP_001171122.1 | 17 | 3920 | |||
NM_006359.2 | NP_006350.1 | 16 | 3946 | |||
XM_006724726.3 | XP_006724789.1 | 17 | 4036 | |||
XM_011531243.2 | XP_011529545.1 | 17 | 3800 | |||
XM_017029223.1 | XP_016884712.1 | 17 | 4053 | |||
XM_017029224.1 | XP_016884713.1 | 17 | 4183 | |||
XM_017029225.1 | XP_016884714.1 | 17 | 3820 | |||
GenBank mRNA | AF030409.1 | - | 16 | 3903 | ||
BC035029.1 | - | 16 | 4004 | 5264865 | ||
BC049169.1 | - | 16 | 3965 | 5312949 | ||
D87743.1 | - | 16 | 3859 |