Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006467.2 | NP_006458.2 | 8 | 868 | ||
XM_005248404.3 | XP_005248461.1 | 8 | 833 | |||
XM_011543100.2 | XP_011541402.1 | 8 | 1101 | |||
XM_011543101.2 | XP_011541403.1 | 8 | 760 | |||
XM_011543102.2 | XP_011541404.1 | 9 | 990 | |||
XM_011543103.2 | XP_011541405.1 | 8 | 887 | |||
XM_011543104.2 | XP_011541406.1 | 8 | 877 | |||
XM_011543105.2 | XP_011541407.1 | 7 | 941 | |||
XM_017008957.1 | XP_016864446.1 | 8 | 1404 | |||
GenBank mRNA | AK295434.1 | - | 8 | 833 | ||
CD657312.1 | - | 5 | 429 | |||
U93868.1 | - | 8 | 829 |