Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006496.3 | NP_006487.1 | 3 | 433 | ||
GenBank mRNA | AF493907.1 | - | 3 | 276 | ||
AK311691.1 | - | 3 | 363 | |||
AK312252.1 | - | 3 | 362 | |||
BC025285.1 | - | 3 | 386 | 5433334 | ||
BP289420.1 | - | 3 | 433 | |||
BT019973.1 | - | 3 | 276 | |||
BT019974.1 | - | 3 | 276 | |||
J03005.1 | - | 3 | 284 | |||
J03198.1 | - | 3 | 338 | |||
J03238.1 | - | 3 | 366 | |||
M27543.1 | - | 3 | 318 |