Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282425.1 | NP_001269354.1 | 4 | 1411 | ||
NM_006572.5 | NP_006563.2 | 4 | 1721 | |||
GenBank mRNA | AK002117.1 | - | 1 | 183 | ||
AK302400.1 | - | 4 | 1411 | |||
BC036756.1 | - | 4 | 1566 | 5548632 |