Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007102.2 | NP_009033.1 | 3 | 532 | ||
GenBank mRNA | U34279.1 | - | 3 | 531 | ||
Z50753.1 | - | 3 | 523 |