Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033583.2 | NP_001028755.2 | 15 | 1480 | ||
NM_001037171.1 | NP_001032248.1 | 16 | 1507 | |||
XM_005274472.2 | XP_005274529.1 | 14 | 1379 | |||
GenBank mRNA | AF132950.1 | - | 14 | 1382 | ||
AF241787.1 | - | 6 | 899 | |||
AK000659.1 | - | 15 | 1466 | |||
AK024337.1 | - | 17 | 2572 | |||
BC136671.1 | - | 15 | 1431 | 9020661 | ||
BC144360.1 | - | 15 | 1431 | 9052881 | ||
BC144370.1 | - | 14 | 1357 | |||
BX648512.1 | - | 16 | 1491 |