Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015270.4 | NP_056085.1 | 22 | 4284 | ||
XM_006719210.3 | XP_006719273.1 | 22 | 5242 | |||
XM_017018743.1 | XP_016874232.1 | 18 | 4446 | |||
GenBank mRNA | AB007882.2 | - | 21 | 4066 | ||
AF250226.1 | - | 21 | 4783 | |||
BC064923.1 | - | 23 | 4206 | |||
BX537664.1 | - | 7 | 2943 |