Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018158.2 | NP_060628.2 | 1 | 383 | ||
GenBank mRNA | AK001486.1 | - | 1 | 383 | ||
AY028435.1 | - | 1 | 359 | |||
BC098302.1 | - | 1 | 378 | 40026925 | ||
BC098358.1 | - | 1 | 378 | 40026922 | ||
BC099711.1 | - | 1 | 378 | 40026923 | ||
BC099739.1 | - | 1 | 376 | 40026926 |