Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020546.2 | NP_065433.2 | 5 | 927 | ||
XM_011513942.2 | XP_011512244.1 | 5 | 874 | |||
GenBank mRNA | AB028983.1 | - | 2 | 248 | ||
AF410885.1 | - | 5 | 886 | |||
AK123013.1 | - | 4 | 828 | |||
AK127036.1 | - | 5 | 875 | |||
AK294555.1 | - | 2 | 383 | |||
AK295531.1 | - | 2 | 267 | |||
AK304041.1 | - | 5 | 762 | |||
BC111743.1 | - | 5 | 865 | 40038765 |