Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC24A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020689.3 | NP_065740.2 | 12 | 1315 | ||
GenBank mRNA | AF169257.2 | - | 12 | 1061 | ||
AF288087.1 | - | 12 | 1315 |