Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128432.2 | NP_001121904.1 | 2 | 354 | ||
NM_001277225.1 | NP_001264154.1 | 2 | 256 | |||
NM_020973.4 | NP_066024.1 | 2 | 354 | |||
NR_102355.1 | 2 | 354 | ||||
NR_102356.1 | 2 | 354 | ||||
NR_102357.1 | 2 | 256 | ||||
GenBank mRNA | AB017913.1 | - | 2 | 303 | ||
AF317840.1 | - | 2 | 252 | |||
AF323990.1 | - | 2 | 301 | |||
AJ278964.1 | - | 2 | 252 | |||
AK222963.1 | - | 2 | 301 | |||
AK292746.1 | - | 2 | 301 | |||
AK298377.1 | - | 2 | 256 | |||
AK313189.1 | - | 2 | 306 | |||
BC029362.1 | - | 2 | 343 | |||
BC070188.1 | - | 2 | 334 | |||
BC101829.1 | - | 2 | 289 | 8069335 | ||
BC109377.1 | - | 2 | 334 | |||
BC136264.1 | - | 2 | 289 | 9020251 | ||
BP383134.1 | - | 354 |