Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277225.1 | NP_001264154.1 | 3 | 403 | ||
NM_020973.4 | NP_066024.1 | 3 | 501 | |||
NR_102355.1 | 3 | 501 | ||||
NR_102357.1 | 3 | 403 | ||||
GenBank mRNA | AB017913.1 | - | 3 | 450 | ||
AF323990.1 | - | 3 | 448 | |||
AJ278964.1 | - | 3 | 399 | |||
AK222963.1 | - | 3 | 448 | |||
AK292746.1 | - | 3 | 448 | |||
AK298377.1 | - | 3 | 403 | |||
AK313189.1 | - | 3 | 453 | |||
BC101829.1 | - | 3 | 436 | 8069335 | ||
BC136264.1 | - | 3 | 436 | 9020251 |