Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128432.2 | NP_001121904.1 | 3 | 956 | ||
NM_001277225.1 | NP_001264154.1 | 5 | 1779 | |||
NM_020973.4 | NP_066024.1 | 5 | 1877 | |||
NR_102355.1 | 5 | 1877 | ||||
NR_102356.1 | 3 | 956 | ||||
NR_102357.1 | 5 | 1779 | ||||
GenBank mRNA | AF323990.1 | - | 5 | 1825 | ||
AK298377.1 | - | 5 | 1780 | |||
BC029362.1 | - | 3 | 947 | |||
BC070188.1 | - | 3 | 937 | |||
BC109377.1 | - | 3 | 937 |