Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPOLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022894.3 | NP_075045.2 | 7, 8 | 853 | ||
XM_005264500.3 | XP_005264557.1 | 7, 8 | 853 | |||
XM_005264501.1 | XP_005264558.1 | 7, 8 | 671 | |||
GenBank mRNA | AF312211.1 | - | 7, 8 | 835 | ||
AJ308101.1 | - | 7, 8 | 604 | |||
AK021867.1 | - | 7, 8 | 789 | |||
AK023544.1 | - | 6, 7 | 602 | |||
AK314663.1 | - | 7, 8 | 817 | |||
AY029162.1 | - | 7, 8 | 613 | |||
BC016738.1 | - | 7, 8 | 824 | |||
BC110906.1 | - | 7, 8 | 792 | |||
BC111701.1 | - | 7, 8 | 727 | 40038554 | ||
BC113747.1 | - | 7, 8 | 727 | 40038563 |