Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001484.2 | NP_001001484.1 | 3 | 594 | ||
NM_001261836.1 | NP_001248765.1 | 2 | 460 | |||
NM_001261837.1 | NP_001248766.1 | 3 | 594 | |||
NM_030664.4 | NP_109589.2 | 3 | 539 | |||
XM_017016927.1 | XP_016872416.1 | 2 | 378 | |||
XM_017016928.1 | XP_016872417.1 | 2 | 554 | |||
XM_017016929.1 | XP_016872418.1 | 2 | 460 | |||
XM_017016930.1 | XP_016872419.1 | 2 | 431 | |||
GenBank mRNA | AF212237.1 | - | 3 | 450 | ||
AK095486.1 | - | 3 | 554 | |||
AK314910.1 | - | 2 | 460 | |||
BC015092.1 | - | 3 | 517 | 4049884 | ||
BC050411.1 | - | 3 | 567 | 6472327 | ||
BX648556.1 | - | 2 | 432 | |||
DA146922.1 | - | 2 | 460 |