Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303484.1 | NP_001290413.1 | 3 | 597 | ||
NM_031481.2 | NP_113669.1 | 4 | 660 | |||
XM_011546149.2 | XP_011544451.1 | 4 | 1589 | |||
XM_011546150.2 | XP_011544452.1 | 4 | 1459 | |||
XM_011546151.2 | XP_011544453.1 | 4 | 848 | |||
XM_011546152.1 | XP_011544454.1 | 4 | 475 | |||
XM_011546153.2 | XP_011544455.1 | 4 | 1470 | |||
XM_011546154.2 | XP_011544456.1 | 4 | 717 | |||
XM_017028968.1 | XP_016884457.1 | 4 | 343 | |||
XM_017028969.1 | XP_016884458.1 | 4 | 2489 | |||
XM_017028970.1 | XP_016884459.1 | 5 | 1980 | |||
XM_017028971.1 | XP_016884460.1 | 5 | 1969 | |||
XM_017028972.1 | XP_016884461.1 | 5 | 1892 | |||
XM_017028973.1 | XP_016884462.1 | 4 | 1917 | |||
XM_017028974.1 | XP_016884463.1 | 5 | 1848 | |||
XM_017028975.1 | XP_016884464.1 | 5 | 1760 | |||
GenBank mRNA | AJ428203.1 | - | 4 | 616 | ||
AK095706.1 | - | 3 | 1582 | |||
AK309110.1 | - | 3 | 444 | |||
AY008285.1 | - | 4 | 616 | |||
BC031644.1 | - | 3 | 552 | 5179038 | ||
CR456578.1 | - | 2 | 246 |