Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303484.1 | NP_001290413.1 | 3, 4 | 610 | ||
NM_031481.2 | NP_113669.1 | 4, 5 | 673 | |||
XM_011546149.2 | XP_011544451.1 | 4, 5 | 1602 | |||
XM_011546150.2 | XP_011544452.1 | 4, 5 | 1472 | |||
XM_011546151.2 | XP_011544453.1 | 4, 5 | 861 | |||
XM_011546152.1 | XP_011544454.1 | 4, 5 | 488 | |||
XM_011546153.2 | XP_011544455.1 | 4, 5 | 1483 | |||
XM_011546154.2 | XP_011544456.1 | 4, 5 | 730 | |||
XM_017028968.1 | XP_016884457.1 | 4, 5 | 356 | |||
XM_017028969.1 | XP_016884458.1 | 4, 5 | 2502 | |||
XM_017028970.1 | XP_016884459.1 | 5, 6 | 1993 | |||
XM_017028971.1 | XP_016884460.1 | 5, 6 | 1982 | |||
XM_017028972.1 | XP_016884461.1 | 5, 6 | 1905 | |||
XM_017028973.1 | XP_016884462.1 | 4, 5 | 1930 | |||
XM_017028974.1 | XP_016884463.1 | 5, 6 | 1861 | |||
XM_017028975.1 | XP_016884464.1 | 5, 6 | 1773 | |||
GenBank mRNA | AJ428203.1 | - | 4, 5 | 629 | ||
AK095706.1 | - | 3, 4 | 1595 | |||
AK309110.1 | - | 3, 4 | 457 | |||
AY008285.1 | - | 4, 5 | 629 | |||
BC031644.1 | - | 3, 4 | 565 | 5179038 | ||
CR456578.1 | - | 2, 3 | 259 |