Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303484.1 | NP_001290413.1 | 10 | 1939 | ||
NM_031481.2 | NP_113669.1 | 11 | 2002 | |||
XM_011546149.2 | XP_011544451.1 | 11 | 2931 | |||
XM_011546150.2 | XP_011544452.1 | 11 | 2801 | |||
XM_011546151.2 | XP_011544453.1 | 11 | 2190 | |||
XM_011546152.1 | XP_011544454.1 | 11 | 1817 | |||
XM_011546153.2 | XP_011544455.1 | 11 | 2812 | |||
XM_011546154.2 | XP_011544456.1 | 11 | 2059 | |||
XM_017028968.1 | XP_016884457.1 | 11 | 1685 | |||
XM_017028969.1 | XP_016884458.1 | 11 | 3831 | |||
XM_017028970.1 | XP_016884459.1 | 12 | 3322 | |||
XM_017028971.1 | XP_016884460.1 | 12 | 3311 | |||
XM_017028972.1 | XP_016884461.1 | 12 | 3234 | |||
XM_017028973.1 | XP_016884462.1 | 11 | 3259 | |||
XM_017028974.1 | XP_016884463.1 | 12 | 3190 | |||
XM_017028975.1 | XP_016884464.1 | 12 | 3102 | |||
GenBank mRNA | AJ428203.1 | - | 11 | 1958 | ||
AY008285.1 | - | 11 | 1958 | |||
BC031644.1 | - | 10 | 1894 | 5179038 |