Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPOLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293627.1 | NP_001280556.1 | 11 | 1202 | ||
NM_001293628.1 | NP_001280557.1 | 11 | 1067 | |||
NM_001293632.1 | NP_001280561.1 | 9 | 1038 | |||
NM_032632.4 | NP_116021.2 | 11 | 1202 | |||
XM_005267281.1 | XP_005267338.1 | 11 | 1198 | |||
XM_005267282.2 | XP_005267339.1 | 11 | 1198 | |||
XM_017020935.1 | XP_016876424.1 | 11 | 1198 | |||
XM_017020936.1 | XP_016876425.1 | 11 | 1198 | |||
GenBank mRNA | AK295020.1 | - | 11 | 1033 | ||
AK302406.1 | - | 12 | 1063 | |||
AK307659.1 | - | 9 | 997 | |||
AK307680.1 | - | 1 | 50 | |||
BC036014.1 | - | 11 | 1197 | 5271668 | ||
BX248301.1 | - | 11 | 1067 | |||
X76770.1 | - | 10 | 961 |