Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001653.2 | NP_001001653.1 | 6 | 749 | ||
NM_052877.4 | NP_443109.2 | 6 | 706 | |||
NM_201542.4 | NP_963836.2 | 6 | 706 | |||
GenBank mRNA | AF521562.1 | - | 5 | 658 | ||
BC010019.2 | - | 6 | 721 | 2905623 | ||
BC010543.1 | - | 5 | 647 |