Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPT2-EGFL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037861.1 | 5 | 859 | |||
GenBank mRNA | AF020544.1 | - | 5 | 1013 | ||
AK290473.1 | - | 5 | 714 | |||
AK296648.1 | - | 4 | 634 | |||
AK307427.1 | - | 3 | 312 |
Gene Symbol: PPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204103.1 | NP_001191032.1 | 5 | 849 | ||
NM_005155.6 | NP_005146.4 | 5 | 859 | |||
NM_138717.2 | NP_619731.2 | 5 | 638 | |||
GenBank mRNA | AF020543.1 | - | 5 | 804 | ||
AL110128.1 | - | 5 | 566 | |||
BC001355.2 | - | 5 | 673 | 3050231 | ||
CR533546.1 | - | 4 | 463 | |||
Y17958.1 | - | 5 | 753 |