Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003487.3 | NP_003478.1 | 6 | 468 | ||
NM_139215.2 | NP_631961.1 | 6 | 477 | |||
XM_011525315.1 | XP_011523617.1 | 6 | 447 | |||
XM_017025181.1 | XP_016880670.1 | 5 | 355 | |||
GenBank mRNA | AK307637.1 | - | 6 | 488 | ||
AK313223.1 | - | 6 | 434 | |||
AK314194.1 | - | 6 | 448 | |||
BC046099.1 | - | 6 | 409 | 6067044 | ||
DB166653.1 | - | 6 | 477 | |||
U51334.1 | - | 6 | 448 | |||
X98893.1 | - | 6 | 399 |