Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003487.3 | NP_003478.1 | 16 | 2106 | ||
NM_139215.2 | NP_631961.1 | 16 | 2115 | |||
XM_011525315.1 | XP_011523617.1 | 16 | 2085 | |||
XM_017025181.1 | XP_016880670.1 | 14 | 1958 | |||
GenBank mRNA | BC039129.1 | - | 2 | 656 | ||
BC046099.1 | - | 16 | 2047 | 6067044 | ||
U51334.1 | - | 16 | 2085 | |||
X98893.1 | - | 16 | 2037 |