Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153343.3 | NP_699174.1 | 4 | 706 | ||
GenBank mRNA | AK057370.1 | - | 4 | 651 | ||
AY358676.1 | - | 4 | 647 | |||
BC035035.2 | - | 4 | 611 | 5259487 |