Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173653.3 | NP_775924.1 | 12 | 1544 | ||
XM_011512703.2 | XP_011511005.1 | 9 | 854 | |||
XM_017006202.1 | XP_016861691.1 | 12 | 1517 | |||
XM_017006203.1 | XP_016861692.1 | 11 | 1302 | |||
GenBank mRNA | AB075486.1 | - | 2 | 45 | ||
AB089794.1 | - | 12 | 1335 | |||
AK092932.1 | - | 6 | 595 | |||
AK315107.1 | - | 12 | 1471 | |||
AL832304.1 | - | 12 | 1409 | |||
AY254100.1 | - | 12 | 1459 | |||
BC035779.1 | - | 12 | 1460 | 5590356 |