Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GANC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301409.1 | NP_001288338.1 | 4, 5 | 902 | ||
NM_198141.2 | NP_937784.2 | 3, 4 | 515 | |||
GenBank mRNA | AF545045.1 | - | 3, 4 | 209 | ||
AF545046.1 | - | 3, 4 | 209 | |||
AK074037.1 | - | 3, 4 | 407 | |||
AK074330.1 | - | 4, 5 | 904 | |||
BC059406.1 | - | 3, 4 | 387 | 30330796 | ||
BC093833.1 | - | 3, 4 | 232 | 7939678 | ||
BC112051.1 | - | 3, 4 | 232 | 8327519 |