Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GANC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198141.2 | NP_937784.2 | 15 | 1985 | ||
GenBank mRNA | AF525397.1 | - | 11 | 1238 | ||
AF545045.1 | - | 15 | 1679 | |||
AF545046.1 | - | 15 | 1679 | |||
AK074037.1 | - | 15 | 1877 | |||
BC059406.1 | - | 15 | 1857 | 30330796 | ||
BC093833.1 | - | 15 | 1702 | 7939678 | ||
BC112051.1 | - | 15 | 1702 | 8327519 |