Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004252.4 | NP_004243.1 | 1 | 432 | ||
GenBank mRNA | AF015926.1 | - | 1 | 409 | ||
AF036241.1 | - | 1 | 367 | |||
AK026581.1 | - | 1 | 415 | |||
AK128474.1 | - | 2 | 381 | |||
BC001443.2 | - | 1 | 385 | 3138881 | ||
BC003361.1 | - | 1 | 402 | 2900516 | ||
BC011777.1 | - | 1 | 401 | 3640162 | ||
BC049220.1 | - | 1 | 408 | |||
BC053350.1 | - | 1 | 392 | 5218414 | ||
BP347679.1 | - | 1 | 420 | |||
CX761192.1 | - | 1 | 432 |