Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006591.2 | NP_006582.1 | 2, 3 | 242 | ||
NR_046410.1 | 2, 3 | 486 | ||||
XM_005273716.1 | XP_005273773.1 | 2, 3 | 190 | |||
XM_011544734.2 | XP_011543036.1 | 2, 3 | 201 | |||
GenBank mRNA | AK302677.1 | - | 2, 3 | 192 | ||
AK316301.1 | - | 2, 3 | 168 | |||
AK316370.1 | - | 2, 3 | 191 | |||
BC032636.1 | - | 2, 3 | 173 | |||
BC058840.1 | - | 2, 3 | 160 | |||
BC092454.1 | - | 2, 3 | 174 | |||
BC108909.1 | - | 2, 3 | 486 | 40010009 | ||
D26018.1 | - | 2, 3 | 188 | |||
DC402448.1 | - | 2, 3 | 243 |