Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301129.1 | NP_001288058.1 | 6 | 597 | ||
NM_021974.4 | NP_068809.1 | 5 | 499 | |||
NR_125371.1 | 5 | 558 | ||||
GenBank mRNA | AK311128.1 | - | 4 | 976 | ||
AK311831.1 | - | 5 | 463 | |||
BC003582.1 | - | 5 | 477 | 3546712 | ||
CR456546.1 | - | 5 | 389 | |||
DA203006.1 | - | 5 | 522 | |||
DA372532.1 | - | 5 | 499 |