Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133489.2 | NP_597996.2 | 10, 11 | 1655 | ||
GenBank mRNA | AF331523.1 | - | 10, 11 | 1655 | ||
AK122981.1 | - | 11, 12 | 1323 | |||
AL050358.1 | - | 4, 5 | 2492 |