Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000342.3 | NP_000333.1 | 18 | 2599 | ||
XM_005257593.4 | XP_005257650.1 | 16 | 2367 | |||
XM_011525129.1 | XP_011523431.1 | 17 | 2536 | |||
GenBank mRNA | AB526447.1 | - | 16 | 2453 | ||
BC096106.3 | - | 17 | 2512 | 40001948 | ||
BC096107.3 | - | 17 | 2512 | 40001951 | ||
BC099628.3 | - | 17 | 2512 | 40001946 | ||
BC099629.3 | - | 17 | 2512 | 40001953 | ||
BC101570.1 | - | 18 | 2536 | 8069076 | ||
BC101574.1 | - | 17 | 2532 | 8069080 | ||
DQ419529.1 | - | 17 | 2518 | |||
GQ981383.1 | - | 17 | 2493 | |||
GQ981384.1 | - | 17 | 2493 | |||
GQ981385.1 | - | 12 | 1395 | |||
GQ981386.1 | - | 17 | 2493 | |||
GQ981387.1 | - | 17 | 2493 | |||
M27819.1 | - | 18 | 2564 | |||
X12609.1 | - | 18 | 2600 |