Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184732.1 | NP_001171661.1 | 4, 5 | 710 | ||
NM_001184733.1 | NP_001171662.1 | 4, 5 | 458 | |||
NM_001184736.1 | NP_001171665.1 | 3, 4 | 373 | |||
NM_004254.3 | NP_004245.2 | 4, 5 | 731 | |||
XM_011545364.1 | XP_011543666.1 | 3, 4 | 306 | |||
GenBank mRNA | AB042505.1 | - | 4, 5 | 713 | ||
AF097491.1 | - | 3, 4 | 586 | |||
AK298341.1 | - | 4, 5 | 428 | |||
AK298351.1 | - | 5, 6 | 683 | |||
AK307850.1 | - | 3, 4 | 338 | |||
AK313187.1 | - | 4, 5 | 704 | |||
BC022387.1 | - | 4, 5 | 725 | 4608542 | ||
BP276255.1 | - | 4, 5 | 428 |