Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184732.1 | NP_001171661.1 | 11 | 1881 | ||
NM_001184733.1 | NP_001171662.1 | 11 | 1629 | |||
NM_001184736.1 | NP_001171665.1 | 10 | 1544 | |||
NM_004254.3 | NP_004245.2 | 11 | 1902 | |||
XM_011545364.1 | XP_011543666.1 | 10 | 1477 | |||
GenBank mRNA | AB042505.1 | - | 11 | 1884 | ||
AB209692.1 | - | 2 | 3626 | |||
AK123243.1 | - | 2 | 2210 | |||
AK298341.1 | - | 11 | 1599 | |||
AK298351.1 | - | 12 | 1854 | |||
BC022387.1 | - | 11 | 1896 | 4608542 |