Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271765.1 | NP_001258694.1 | 7 | 1822 | ||
NM_004695.3 | NP_004686.1 | 7 | 1756 | |||
XM_005257790.1 | XP_005257847.1 | 6 | 1698 | |||
XM_011525462.1 | XP_011523764.1 | 6 | 1646 | |||
XM_017025293.1 | XP_016880782.1 | 6 | 3483 | |||
GenBank mRNA | AK000416.1 | - | 7 | 1822 | ||
AK092512.1 | - | 6 | 1865 | |||
BC009684.1 | - | 7 | 1737 | 3895881 | ||
BC033611.1 | - | 6 | 1646 | 5218712 | ||
U59299.1 | - | 5 | 1510 |