Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005073.3 | NP_005064.1 | 13 | 1034 | ||
GenBank mRNA | AF043233.1 | - | 13 | 1013 | ||
AY634368.1 | - | 13 | 1013 | |||
BC096328.3 | - | 13 | 988 | 40008279 | ||
BC096329.3 | - | 13 | 988 | 40008280 | ||
EU544224.1 | - | 12 | 957 | |||
U13173.1 | - | 13 | 1013 | |||
U21936.1 | - | 13 | 1013 |