Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032940.2 | NP_116558.1 | 6, 7 | 529 | ||
GenBank mRNA | AF008443.1 | - | 6, 7 | 443 | ||
AJ224143.1 | - | 6, 7 | 482 | |||
AJ224144.1 | - | 6, 7 | 482 | |||
AK293112.1 | - | 6, 7 | 487 | |||
AK300124.1 | - | 500 | ||||
BC000409.2 | - | 6, 7 | 460 | 2960943 | ||
BC003159.1 | - | 6, 7 | 501 | 2957867 | ||
BC028157.1 | - | 6, 7 | 485 | 5240953 | ||
BI458170.1 | - | 6, 7 | 537 | |||
CR542041.1 | - | 6, 7 | 443 | |||
J05448.1 | - | 6, 7 | 484 |