Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DGKA SIRNA Assays ›
Gene Symbol:
DGKA
Gene Name:
diacylglycerol kinase alpha
Gene Aliases:
DAGK, DAGK1, DGK-alpha
Chromosome Location:
Chr.12: 55931076 - 55954023 on Build GRCh38
UniGene ID:
Hs.524488
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
120
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DGKA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001345.4
NP_001336.2
18
1733
NM_201444.2
NP_958852.1
18
1778
NM_201445.1
NP_958853.1
19
1746
NM_201554.1
NP_963848.1
18
1646
XM_005268688.2
XP_005268745.1
18
1675
XM_005268689.2
XP_005268746.1
18
1610
XM_005268690.2
XP_005268747.1
16
1430
XM_011537991.2
XP_011536293.1
19
1843
XM_011537993.2
XP_011536295.1
18
1594
XM_011537995.2
XP_011536297.1
18
1605
XM_017018900.1
XP_016874389.1
17
2029
XM_017018901.1
XP_016874390.1
17
2173
XM_017018902.1
XP_016874391.1
17
1995
XM_017018903.1
XP_016874392.1
18
2197
XM_017018904.1
XP_016874393.1
18
2065
XM_017018905.1
XP_016874394.1
19
2474
XM_017018906.1
XP_016874395.1
17
2106
XM_017018907.1
XP_016874396.1
17
1929
XM_017018908.1
XP_016874397.1
18
1813
XM_017018909.1
XP_016874398.1
17
1564
GenBank mRNA
AF064767.1
-
4
335
AF064769.1
-
18
1655
AF064770.1
-
18
1723
AF064771.1
-
18
1691
AK122973.1
-
16
3084
AK127093.1
-
11
2543
AK226134.1
-
13
3688
AK307685.1
-
17
1490
AY930112.1
-
17
1584
BC023523.2
-
19
1669
4304187
BC031870.2
-
18
1673
4819985
X62535.1
-
17
1572
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidic acid biosynthetic process
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
platelet activation
intracellular signal transduction
diacylglycerol kinase activity
calcium ion binding
ATP binding
phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-4h6qx:80/100.66.76.46:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline